Hesperetin ≥97.0% (by HPLC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: 3',5,7-Trihydroxy-4'-methoxyflavanone, 4'-Methoxy-3',5,7-trihydroxyflavanone, (S)-5,7-Dihydroxy-2-(3-hydroxy-4-methoxyphenyl)chroman-4-one, (±)-Hesperetin

H0721-25G H0721-5G
TCIAH0721-25GEA 188 EUR
TCIAH0721-25G TCIAH0721-5G
Hesperetin ≥97.0% (by HPLC, titration analysis)
Hesperetin
Formula: C₁₆H₁₄O₆
MW: 302,30 g/mol
Número MDL: MFCD00075646
Núm. CAS: 520-33-2
EINECS: 208-290-2
Índice Merck: 14,04670
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fffd19474f8747eab3b4b959581c959525e1bfbd&vid=fffd19474f8747eab3b4b959581c959525e1bfbd&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 97.0 %
Melting point 227.0 to 233.0 deg-C
Solubility in hot Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR