4-(N,N-Diphenylamino)benzaldehyde ≥98.0% (by GC, total nitrogen)

Proveedor: TCI

Sinónimos: 4-Diphenylaminobenzaldehyde

D2172-25G D2172-5G
TCIAD2172-25GEA 261 EUR
TCIAD2172-25G TCIAD2172-5G
4-(N,N-Diphenylamino)benzaldehyde ≥98.0% (by GC, total nitrogen)
4-(N,N-Diphenylamino)benzaldehyde
Formula: C₁₉H₁₅NO
MW: 273,33 g/mol
Número MDL: MFCD00145131
Núm. CAS: 4181-05-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=816fc95fa1d7ddebf0f10e511f10c26636eab11f&vid=816fc95fa1d7ddebf0f10e511f10c26636eab11f&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance Light yellow to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (with Total Nitrogen) min. 98.0 %
Melting point 131.0 to 135.0 deg-C
Solubility in hot Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR