4-(Phenylazo)diphenylamine ≥98.0% (by titrimetric analysis)

Proveedor: TCI
Atención

P0146-25G P0146-5G
TCIAP0146-25GEA 119 EUR
TCIAP0146-25G TCIAP0146-5G
4-(Phenylazo)diphenylamine ≥98.0% (by titrimetric analysis)
4-(Phenylazo)diphenylamine
Formula: C₁₈H₁₅N₃
Número MDL: MFCD00003023
Núm. CAS: 101-75-7
EINECS: 202-972-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6316c51eed7595bab7e77e91ade214cd42afaba4&vid=6316c51eed7595bab7e77e91ade214cd42afaba4&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance Light yellow to Brown powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 86.0 to 90.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR