Benzophenone-p-toluenesulphonylhydrazone ≥97%

Proveedor: Thermo Scientific

H53486.06 H53486.14
H53486.06EA 43.9 EUR
H53486.06 H53486.14
Benzophenone-p-toluenesulphonylhydrazone ≥97%
Benzophenone-p-toluenesulphonylhydrazone

Purity: 97% CAS No.: 4545-20-4 Molecular Formula: C20H18N2O2S Formula Weight: 350.44 Melting Point: 178-182° MDL No.: MFCD00381345

Formula: C₂₀H₁₈N₂O₂S
MW: 350,44 g/mol
Pto. de fusión: 178…182 °C
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00381345
Núm. CAS: 4545-20-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR