N,N-Diethylnicotinamide ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Peligro

D0514-25G D0514-500G
TCIAD0514-25GEA 24 EUR
TCIAD0514-25G TCIAD0514-500G
N,N-Diethylnicotinamide ≥98.0% (by GC, titration analysis)
N,N-Diethylnicotinamide
Formula: C₁₀H₁₄N₂O
Número MDL: MFCD00006386
Núm. CAS: 59-26-7
EINECS: 200-418-5
UN: 2811
ADR: 6.1,III
Índice Merck: 14,06543
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7b96d5afa69068b4d6556202e6953a8ddad3a3bf&vid=7b96d5afa69068b4d6556202e6953a8ddad3a3bf&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White or Colorless to Light orange to Yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR