2-Chloro-4'-phenylacetophenone ≥99.0% (by GC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: 1-([1,1-Biphenyl]-4-yl)-2-chloroethanone

C0252-25G C0252-5G
TCIAC0252-25GEA 167 EUR
TCIAC0252-25G TCIAC0252-5G
2-Chloro-4'-phenylacetophenone ≥99.0% (by GC, titration analysis)
2-Chloro-4'-phenylacetophenone
Formula: C₁₄H₁₁ClO
MW: 230,6895 g/mol
Número MDL: MFCD00058937
Núm. CAS: 635-84-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eb46687f847863cb75dbdf4f55d283e93b4f7faa&vid=eb46687f847863cb75dbdf4f55d283e93b4f7faa&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Light yellow powder to crystal
Purity (GC) min. 99.0 %
Purity (Argentmetric Titration) min. 99.0 %
Melting point 126.0 to 131.0 deg-C
Solubility in hot Acetonitrile almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR