Palmitic anhydride ≥96.0% (by GC, titration analysis)
Proveedor: TCI
|
Peligro
|
P0008-25G
P0008-500G
TCIAP0008-25GEA
49
EUR
TCIAP0008-25G
TCIAP0008-500G
Palmitic anhydride ≥96.0% (by GC, titration analysis)
Palmitic anhydride
Formula:
C₃₂H₆₂O₃ MW: 494.8329 g/mol |
Número MDL:
MFCD00008992 Núm. CAS: 623-65-4 EINECS: 210-805-0 UN: 3261 ADR: 8 III |
|
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 96.0 % |
Purity (Morpholine Method) | min. 96.0 % |
Melting point | 62.0 to 65.0 deg-C |
Solubility in Toluene | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...