Trimethylamine-N-oxide, anhidro ≥95.0% (by titrimetric analysis)

Proveedor: TCI
Atención

Sinónimos: TMANO

T1362-1G T1362-5G
TCIAT1362-1GEA 57 EUR
TCIAT1362-1G TCIAT1362-5G
Trimethylamine-N-oxide, anhidro ≥95.0% (by titrimetric analysis)
Trimethylamine-N-oxide
Formula: C₃H₉NO
MW: 75,11 g/mol
Número MDL: MFCD00002048
Núm. CAS: 1184-78-7
EINECS: 214-675-6
Índice Merck: 14,09711
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a81ac26327988ec2cb4d8145d1bf86efcc67b49c&vid=9c7d5bc5a8e67667c6c6141f1a38c90cb0bd81a0&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystaline
Purity (Nonaqueous Titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR