1-Cetilpiridinio bromuro, hidrato ≥96.0% (by HPLC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: Bromuro de 1-cetilpiridinio, Bromuro de cetilpiridinio

H0162-25G H0162-500G
TCIAH0162-25GEA 18 EUR
TCIAH0162-25G TCIAH0162-500G
1-Cetilpiridinio bromuro, hidrato ≥96.0% (by HPLC, titration analysis)
1-Cetilpiridinio bromuro
Formula: C₂₁H₃₈BrN·xH₂O
MW: 384.44 g/mol (anhy)
Número MDL: MFCD00011988
Núm. CAS: 140-72-7
EINECS: 205-428-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=550fb2b4bcd0d9b069e89d945ebd0368f4122caf&vid=24b47e802e3e29f87b7b73aa2ad6191fe3b0d1ff&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 96.0 area%
Purity (Nonaqueous Titration) min. 96.0 % (calcd.on anh.substance)
Solubility in Methanol almost transparency
Solubility in hot Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR