2-Methylhippuric acid ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI

Sinónimos: 2-(2-Methylbenzamido)acetic acid

T0719-25G T0719-5G
TCIAT0719-25GEA 236 EUR
TCIAT0719-25G TCIAT0719-5G
2-Methylhippuric acid ≥98.0% (by HPLC, titration analysis)
2-Methylhippuric acid
Formula: C₁₀H₁₁NO₃
Número MDL: MFCD00050991
Núm. CAS: 42013-20-7
EINECS: 255-622-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5f31d2ebf60b70f0a4c10160e42f6de9e77b9224&vid=5f31d2ebf60b70f0a4c10160e42f6de9e77b9224&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 161.0 to 165.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR