4-Methylhippuric acid ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI

T0313-25G T0313-5G
TCIAT0313-25GEA 80 EUR
TCIAT0313-25G TCIAT0313-5G
4-Methylhippuric acid ≥98.0% (by HPLC, titration analysis)
4-Methylhippuric acid
Formula: C₁₀H₁₁NO₃
Número MDL: MFCD00020449
Núm. CAS: 27115-50-0
EINECS: 248-231-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eab5693e39dac0b3b9ea1e91d8363064fbb5e804&vid=eab5693e39dac0b3b9ea1e91d8363064fbb5e804&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 161.0 to 166.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR