Aluminium perchlorate nonahydrate, grado reactivo
Proveedor: Thermo Fisher Scientific
|
Peligro
|
11659.22
11659.36
11659.22EA
59.7
EUR
11659.22
11659.36
Aluminium perchlorate nonahydrate, grado reactivo
Aluminium perchlorate nonahydrate
CAS No.: 81029-06-3
Molecular Formula: Al(ClO4)3.9H2O
Formula Weight: 487.46 (325.33anhy)
Physical Form: Crystalline
Sensitivity: Hygroscopic
Melting Point: 82°
UN No.: 1481
MDL No.: MFCD00149130
Formula:
AlCl₃O₁₂·9H₂O Pto. de fusión: 82 °C Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00149130 Núm. CAS: 81029-06-3 EINECS: 238-437-6 UN: 1481 ADR: 5.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...