trans-m-Coumaric acid ≥99.0% (by GC, titration analysis)

Proveedor: TCI
Atención

C0655-25G C0655-5G
TCIAC0655-25GEA 47 EUR
TCIAC0655-25G TCIAC0655-5G
trans-m-Coumaric acid ≥99.0% (by GC, titration analysis)
trans-m-Coumaric acid
Formula: C₉H₈O₃
Número MDL: MFCD00004390
Núm. CAS: 14755-02-3
EINECS: 209-615-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=63679b6f7fdb861a469d702e2115655986a7f939&vid=26e24dc752c27cbdacc993d8308fd91677346d36&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Purity (Neutralization titration) min. 99.0 %
Melting point 194.0 to 198.0 deg-C
Solubility in Methanol almost transparency

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR