5-Bromooxindol ≥97.0% (by GC, total nitrogen)
Proveedor: TCI
|
Atención
|
Sinónimos:
5-Bromo-1,3-dihydro-2H-indol-2-one, 5-Bromoindolin-2-one, 5-Bromo-2-oxindole
B3741-5G
B3741-1G
TCIAB3741-5GEA
120
EUR
TCIAB3741-5G
TCIAB3741-1G
5-Bromooxindol ≥97.0% (by GC, total nitrogen)
5-Bromooxindol
Formula:
C₈H₆BrNO MW: 212,05 g/mol |
Número MDL:
MFCD00456998 Núm. CAS: 20870-78-4 |
Resultados test especificaciones
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (with Total Nitrogen) | min. 97.0 % |
Melting point | 220.0 to 224.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...