Diacetato de 4,4'-isopropilidendifenilo ≥98.0% (por GC)
Proveedor: TCI
Sinónimos:
Bisphenol A diacetate, 2,2-Bis(4-acetoxyphenyl)propane, 4,4'-Isopropylidenediphenol diacetate
B1286-25G
B1286-500G
TCIAB1286-25GEA
26
EUR
TCIAB1286-25G
TCIAB1286-500G
Diacetato de 4,4'-isopropilidendifenilo ≥98.0% (por GC)
Diacetato de 4,4'-isopropilidendifenilo
Formula:
C₁₉H₂₀O₄ Pto. Ebullición: 215°C |
Número MDL:
MFCD00026194 Núm. CAS: 10192-62-8 EINECS: 233-472-3 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 92.0 to 95.0 deg-C |
Solubility in hot Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...