Capillary columns, TraceGOLD™ TG-35MS AMINE
Proveedor: Thermo Fisher Scientific
26092-3350 26092-2230 26092-2860 26092-2960 26092-2970 26092-2980
555-8701EA 1040 EUR
555-8701 555-8708 555-8705 555-8704 555-8703 555-8702
Capillary columns, TraceGOLD™ TG-35MS AMINE
Columnas de cromatografía
Chemically altered tubing surface reduces tailing and eliminates the need for column priming.
  • Midpolarity phase, 35% diphenyl or 65% dimethyl polysiloxane
  • Developed for analysis of active basic compounds without derivatisation
  • Allows analysis of neutral compounds and adsorptive compounds with oxygen groups susceptible to hydrogen bonding
  • Low bleed at maximum operating temperature