Safety shoes for ladies, lace-up, Steelite™ LADIES FW39
Proveedor: Portwest
FW39BKR39 FW39BKR40 FW39BKR42 FW39BKR38 FW39BKR37 FW39BKR36 FW39BKR41 FW39BPR36 FW39BPR37 FW39BPR40 FW39BPR41 FW39BPR42 FW39BPR43 FW39BPR38 FW39BPR39
114-4394EA 64.9 EUR
114-4394 114-4395 114-4397 114-4393 114-4392 114-4391 114-4396 PRTWFW39BPR36 PRTWFW39BPR37 PRTWFW39BPR40 PRTWFW39BPR41 PRTWFW39BPR42 PRTWFW39BPR43 PRTWFW39BPR38 PRTWFW39BPR39
Safety shoes for ladies, lace-up, Steelite™ LADIES FW39
Zapatos y botas
Anti-static, low cut trainer for ladies made from split leather upper plus a dual-density EVA/rubber sole, resistant to oil and heat (300 °C).
  • Shock-absorbent heel
  • Protective steel toecap (200 J)
  • Puncture resistant steel midsole
Colour: Black.
Colour: Black
Certificaciones: EN 20345:2011 + A1:2007; S1P, SRC, HRO.