5-Amino-1-phenyl-1H-pyrazole ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: 1-Phenyl-1H-pyrazol-5-amine, 5-Amino-1-phenylpyrazole

A0174-25G A0174-5G
TCIAA0174-25GEA 409 EUR
TCIAA0174-25G TCIAA0174-5G
5-Amino-1-phenyl-1H-pyrazole ≥98.0% (by GC, titration analysis)
5-Amino-1-phenyl-1H-pyrazole
Formula: C₉H₉N₃
MW: 159,1900 g/mol
Número MDL: MFCD00020734
Núm. CAS: 826-85-7
EINECS: 212-563-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=38f575f7a004a27e774f31c13e1032b306427063&vid=38f575f7a004a27e774f31c13e1032b306427063&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 42.0 to 46.0 deg-C
Solubility in Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR