Methyl arachidonate
Proveedor: Cayman Chemical
90014-100
90014-25
90014-50
CAYM90014-100EA
189
EUR
CAYM90014-100
CAYM90014-25
CAYM90014-50
Methyl arachidonate
Methyl arachidonate
Formula:
C₂₁H₃₄O₂ |
Número MDL:
MFCD00016775 Núm. CAS: 2566-89-4 EINECS: 219-900-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...