1-(Chloromethyl)benzotriazole ≥98.0% (by titrimetric analysis)
Proveedor: TCI
|
Peligro
|
C1526-25G
C1526-5G
TCIAC1526-25GEA
208
EUR
TCIAC1526-25G
TCIAC1526-5G
1-(Chloromethyl)benzotriazole ≥98.0% (by titrimetric analysis)
1-(Chloromethyl)benzotriazole
Formula:
C₇H₆ClN₃ |
Número MDL:
MFCD00216649 Núm. CAS: 54187-96-1 UN: 3261 ADR: 8,II |
|
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (Argentmetric Titration) | min. 98.0 % |
Melting point | 136.0 to 139.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...