Acetyl iodide ≥95.0% (by titrimetric analysis)
Proveedor: TCI
A0095-10G
A0095-25G
TCIAA0095-10GEA
164
EUR
TCIAA0095-10G
TCIAA0095-25G
Acetyl iodide ≥95.0% (by titrimetric analysis)
Acetyl iodide
Resultados test especificaciones
Appearance | Red to Dark red to Brown clear liquid to cloudy liquid |
Purity (Potassium iodate Method) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...