(±)-2-Aminooctanoic acid ≥98.0% (by titrimetric analysis)
Proveedor: TCI
A0382-25G
A0382-5G
TCIAA0382-25GEA
187
EUR
TCIAA0382-25G
TCIAA0382-5G
(±)-2-Aminooctanoic acid ≥98.0% (by titrimetric analysis)
(±)-2-Aminooctanoic acid
Formula:
C₈H₁₇NO₂ |
Número MDL:
MFCD00008102 Núm. CAS: 644-90-6 EINECS: 211-424-2 |
|
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Solubility in hot dilute HCl | very faint turbidity |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...