2-Amino-1-naphthalenesulphonic acid ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: 2-Aminonaphthalene-1-sulphonic acid

A0343-25G A0343-500G
TCIAA0343-25GEA 30 EUR
TCIAA0343-25G TCIAA0343-500G
2-Amino-1-naphthalenesulphonic acid ≥98.0% (by HPLC, titration analysis)
2-Amino-1-naphthalenesulphonic acid
Formula: C₁₀H₉NO₃S
MW: 223,25 g/mol
Número MDL: MFCD00003988
Núm. CAS: 81-16-3
EINECS: 201-331-5
Índice Merck: 14,06406
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dc78fbd8169dd3fe82078b8acc4d889fc801d2ef&vid=dc78fbd8169dd3fe82078b8acc4d889fc801d2ef&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR