Benzyl (R)-(-)-glycidyl ether ≥98.0% (por GC)

Proveedor: TCI
Atención

Sinónimos: (R)-2-((Benzyloxy)methyl)oxirane

B2238-1G B2238-25G B2238-5G
TCIAB2238-1GEA 16 EUR
TCIAB2238-1G TCIAB2238-25G TCIAB2238-5G
Benzyl (R)-(-)-glycidyl ether ≥98.0% (por GC)
Benzyl (R)-(-)-glycidyl ether
Formula: C₁₀H₁₂O₂
MW: 164,20 g/mol
Número MDL: MFCD00068409
Núm. CAS: 14618-80-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6677e4a55aa388780d491d5600fbf2e8e9a0644&vid=f6677e4a55aa388780d491d5600fbf2e8e9a0644&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Specific rotation [a]20/D -13.0 to -18.0 deg (neat)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR