2-Fluoro-1-methylpyridinium-p-toluenesulphonate ≥98.0% (by titrimetric analysis) fluorinating reagent
Proveedor: TCI
|
Atención
|
Sinónimos:
2-Fluoro-1-methylpyridin-1-ium 4-methylbenzenesulphonate
F0225-25G
F0225-5G
TCIAF0225-25GEA
170
EUR
TCIAF0225-25G
TCIAF0225-5G
2-Fluoro-1-methylpyridinium-p-toluenesulphonate ≥98.0% (by titrimetric analysis) fluorinating reagent
2-Fluoro-1-methylpyridinium-p-toluenesulphonate
Formula:
C₁₃H₁₄FNO₃S MW: 283,32 g/mol |
Número MDL:
MFCD00011983 Núm. CAS: 58086-67-2 EINECS: 261-108-3 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Solubility in Water | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...