(Methylsulphonyl)methanesulphonyl chloride ≥98%
Proveedor: Thermo Scientific
|
Peligro
|
H50385.03
H50385.06
H50385.03EA
123
EUR
H50385.03
H50385.06
(Methylsulphonyl)methanesulphonyl chloride ≥98%
(Methylsulphonyl)methanesulphonyl chloride
Purity: 98%
CAS No.: 22317-89-1
Molecular Formula: C2H5ClO4S2
Formula Weight: 192.63
Melting Point: 110°
UN No.: 3261
MDL No.: MFCD00052809
Formula:
C₂H₅ClO₄S₂ MW: 192,63 g/mol Pto. de fusión: 110 °C Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00052809 Núm. CAS: 22317-89-1 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...