Dimethyl-1,4-cyclohexanedione-2,5-dicarboxylate ≥98.0% (by HPLC)

Proveedor: TCI

Sinónimos: Dimethyl 2,5-dioxocyclohexane-1,4-dicarboxylate

C1343-25G C1343-500G
TCIAC1343-25GEA 18 EUR
TCIAC1343-25G TCIAC1343-500G
Dimethyl-1,4-cyclohexanedione-2,5-dicarboxylate ≥98.0% (by HPLC)
Dimethyl-1,4-cyclohexanedione-2,5-dicarboxylate
Formula: C₁₀H₁₂O₆
MW: 228,2 g/mol
Número MDL: MFCD00001607
Núm. CAS: 6289-46-9
EINECS: 228-528-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0d809fc199a306d7eb3f71f99036d83d88ac13e9&vid=0d809fc199a306d7eb3f71f99036d83d88ac13e9&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Light yellow to Green powder to crystaline
Purity (GC) min. 95.0 %
Melting point 153.0 to 157.0 deg-C
Solubility in hot Pyridine very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR