Tetrabutylammonium tetraphenylborate ≥98.0% (by HPLC, total nitrogen)
Proveedor: TCI
|
Atención
|
Sinónimos:
Tetrabutylammonium Tetraphenylborate
T1480-1G
T1480-5G
TCIAT1480-1GEA
23
EUR
TCIAT1480-1G
TCIAT1480-5G
Tetrabutylammonium tetraphenylborate ≥98.0% (by HPLC, total nitrogen)
Tetrabutylammonium tetraphenylborate
Formula:
C₄₀H₅₆BN MW: 561,6904 g/mol |
Número MDL:
MFCD00011749 Núm. CAS: 15522-59-5 EINECS: 239-562-9 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 233.0 to 237.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...