Olive oil, puro, virgen
Proveedor: Thermo Fisher Scientific
416540250
416545000
416540025
ACRO416540250EA
33.2
EUR
ACRO416540250
ACRO416545000
ACRO416540025
Olive oil, puro, virgen
Olive oil
Densidad:
0.9135 g/cm³ Pto. de inflamación: 225 °C Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00131764 Núm. CAS: 8001-25-0 EINECS: 232-277-0 |
Resultados test especificaciones
Appearance | Clear yellow to yellow-green Oily liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4670 to 1.4710 (20°C, 589 nm) |
Water | ≤0.1 % |
Absorbance | ≤0.2 (270nm, 1g/100ml cyclohexane) |
Composition | ≤0.4 % (Eicosenoic acid) |
Acid value | ≤2.0 mg KOH/g |
Peroxide value | ≤20.0 meq O2/kg |
Unsaponifiable matter | ≤1.5 % |
Sesame oil | Complies |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...