4,4-Dimethyl-3-thiosemicarbazide ≥98.0% (by titrimetric analysis)

Proveedor: TCI
Peligro

D2096-1G D2096-5G
TCIAD2096-1GEA 33 EUR
TCIAD2096-1G TCIAD2096-5G
4,4-Dimethyl-3-thiosemicarbazide ≥98.0% (by titrimetric analysis)
4,4-Dimethyl-3-thiosemicarbazide
Formula: C₃H₉N₃S
MW: 119,1887 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD00041308
Núm. CAS: 6926-58-5
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4b1bfa607720e2780f56c1ad70aebe41c7cb49e0&vid=4b1bfa607720e2780f56c1ad70aebe41c7cb49e0&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR