4-Isopropyl-3-thiosemicarbazide
Proveedor: TCI
|
Atención
|
Sinónimos:
N-Isopropylhydrazinecarbothioamide
I0418-1G
I0418-5G
TCIAI0418-1GEA
40
EUR
TCIAI0418-1G
TCIAI0418-5G
4-Isopropyl-3-thiosemicarbazide
4-Isopropyl-3-thiosemicarbazide
Formula:
C₄H₁₁N₃S MW: 133,22 g/mol |
Número MDL:
MFCD00025144 Núm. CAS: 13431-36-2 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 96.0 % |
Melting point | 94.0 to 98.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...