BRP Indicator solution Reag. Ph. Eur. 1013000
Proveedor: VWR Chemicals
87799.180EA
55.5
EUR
87799.180
87799.180_T
BRP Indicator solution Reag. Ph. Eur. 1013000
BRP Indicator solution
Solution of Bromothymol Blue R, Methyl Red R and Phenolphtthalein R in alcohol