1,2-Dinitrobenceno Reag. Ph. Eur. 1031201

Proveedor: VWR Chemicals

87818.290EA 240 EUR
87818.290 87818.290_T
1,2-Dinitrobenceno Reag. Ph. Eur. 1031201
1,2-Dinitrobenceno
Formula: C₆H₄N₂O₄
Número MDL: MFCD00007093
Núm. CAS: 528-29-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR