HPLC columns, Acclaim™ RSLC PolarAdvantage II
Proveedor: Thermo Fisher Scientific
068989 068990 071401 071402 071607 074814
555-7123EA 879 EUR
555-7123 555-7122 555-7215 555-7217 555-7221 555-7510
HPLC columns, Acclaim™ RSLC PolarAdvantage II
Columnas de cromatografía
The Acclaim PA2 column is specifically designed to withstand high pH conditions, for the separation of both basic and acidic analytes.
  • Novel polar-embedded column chemistry for hydrolytic stability
  • Low bleed for MS compatibility
  • Compatible with 100% aqueous mobile phase