HPLC columns, Acclaim™ RSLC Carbonyl
Proveedor: Thermo Fisher Scientific
077972 077973 077974
555-7545EA 1090 EUR
555-7545 555-7546 555-7547
HPLC columns, Acclaim™ RSLC Carbonyl
Columnas de cromatografía
These silica based carbonyl columns provides ideal selectivity for baseline resolution and separation of DNPH derivatives of aldehydes and ketones.