Filter cartridges, captiva
Proveedor: Agilent
A500401000 A5307045 A5300002
HEWLA500401000EA 286 EUR
HEWLA500401000 HEWLA5307045 HEWLA5300002
Filter cartridges, captiva
Filtros Cartuchos de filtros
Captiva filter cartridges with standard SPE format which are ideal for LC/MS samples.
  • Protect HPLC columns from clogging to reduce instrument downtime
  • Clean and clear filtrates offer improved sensitivity
  • Rigorous quality control eliminates nonspecific binding
  • Fast and reliable processing improves productivity
  • High analyte recovery with simple robust methods allows faster method development
Nondrip (3 ml) lipid cartridges are designed for workflows using a vacuum manifold and effectively remove ion suppressing phospholipids from biological samples. 0,2 μm pore size filters can be used with either methanol or acetonitrile.