Temperature sensors for IKA digital dry block heaters
Proveedor: IKA
3516800 4480600
442-0584EA 174 EUR
442-0584 460-1041
Temperature sensors for IKA digital dry block heaters
Pruebas de temperatura
For use with RCT basic, RET basic and C-MAG HS digital series.
PT1000 (DIN IEC 751 Class A)