Rain jackets, Dortmund 4820
Proveedor: SIOEN
4820/Y58/1XL 4820/Y58/2XL 4820/Y58/3XL 4820/Y58/L 4820/Y58/M 4820/Y58/S 4820/A41/1XL 4820/A41/2XL 4820/A41/3XL 4820/A41/L 4820/A41/M 4820/A41/S 4820/B90/1XL 4820/B90/2XL 4820/B90/3XL 4820/B90/L 4820/B90/M 4820/B90/S
SIOE4820/Y58/1XLEA 72.9 EUR
SIOE4820/Y58/1XL SIOE4820/Y58/2XL SIOE4820/Y58/3XL SIOE4820/Y58/L SIOE4820/Y58/M SIOE4820/Y58/S SIOE4820/A41/1XL SIOE4820/A41/2XL SIOE4820/A41/3XL SIOE4820/A41/L SIOE4820/A41/M SIOE4820/A41/S SIOE4820/B90/1XL SIOE4820/B90/2XL SIOE4820/B90/3XL SIOE4820/B90/L SIOE4820/B90/M SIOE4820/B90/S
Rain jackets, Dortmund 4820
Ropa de trabajo
Water- and windproof jacket made of Flexothane® Classic, a polyamide knitting with PU coating. The fabric is lightweight, noiseless and provides extended durability.
  • High frequency welded seams
  • Straight collar with foldaway hood
  • Zip closure under flap with press studs
  • Two patched pockets
  • Raglan sleeves, elasticated wristband with narrowing by press studs
  • Underarm ventilation
Back length: 85 cm
Weight: 180 g/m²
Certificaciones: ENV 343 (3-2)
Order Now
ESPECIFICACIONES
Modelo 4820 Dortmund
Environmentally Preferable
Aplicaciones de vestimenta General Purpose
Risk area Rain (waterproof)
Género Unisex
Sleeve or leg length Long
Tamaño de cierre Con botones a presión
Tela Polyamide knitting/PU coating
Peso 180 g/m²