Overalls, standard, C802
Proveedor: Portwest
C802NARXXXL C802NARXXL C802NARXL C802NAR5XL C802NARM C802NARL C802NAR4XL C802NARS C802BKRM C802BKRXL C802BKRS C802BKRXXL C802BKRL
PRTWC802NARXXXLEA 40 EUR
PRTWC802NARXXXL PRTWC802NARXXL PRTWC802NARXL PRTWC802NAR5XL PRTWC802NARM PRTWC802NARL PRTWC802NAR4XL PRTWC802NARS PRTWC802BKRM PRTWC802BKRXL PRTWC802BKRS PRTWC802BKRXXL PRTWC802BKRL
Overalls, standard, C802
Ropa de trabajo
These standard overalls are made of Kingsmill fabric consisting of 65% polyester and 35% cotton. The material is fully pre-shrunk and has superior dye retention.
  • One chest, one rule and two side pockets
  • Back elastication provides all day comfort
  • Concealed zip fastening
Weight: 245 g/m²
Order Now
ESPECIFICACIONES
Modelo Standard C802
Environmentally Preferable
Aplicaciones de vestimenta General Purpose,Industrial
Risk area
Género Unisex
Sleeve or leg length Long
Tamaño de cierre Con cremallera
Tela Kingsmill fabric consisting of 65% polyester and 35% cotton
Peso 245 g/m²