Chemical resistant overalls, type 5/6, BizTex® Microcool, ST50
Proveedor: Portwest
ST50WHRL ST50WHRM ST50WHRXL ST50WHRXXL ST50WHRXXXL
PRTWST50WHRLEA 383 EUR
PRTWST50WHRL PRTWST50WHRM PRTWST50WHRXL PRTWST50WHRXXL PRTWST50WHRXXXL
Chemical resistant overalls, type 5/6, BizTex® Microcool, ST50
Prendas desechables
This anti-static overall made of BizTex® fabric (PP/PE) combined with SMS (panel at hood, back and upper legs) offers enhanced comfort and protection.
  • Breathable comfort panel made of SMS
  • Provides excellent chemical barrier and lint free properties
Weight: 63 g/m² (PP/PE); 55 g/m² (SMS)
Certificaciones: EN 1149-5, EN 13034 (type 6), EN ISO 13982 (Type 5), EN 1073-2; Cat. III