Barrier tips, ART™
Proveedor: Thermo Fisher Scientific
2069-HR 2139-HR 2149E-HR 2149P-HR 2179-HR 2069-RT 2070-HR 2070-RT 2139-RT 2149P-RT 2179-RT
732-1145EA 0 EUR
732-1145 732-1139 732-1141 732-1142 732-1146 732-2392 732-2395 732-2396 732-2402 732-2404 732-2408
Barrier tips, ART™
Puntas de pipetas
These sterile barrier tips are designed to fit virtually any pipette and application. The self-sealing barrier provides 100% protection against aerosols and liquid contamination.
  • Free of RNase, DNase, DNA, ATP, Bioburden, Endotoxin, Pyrogen and PCR Inhibitors
  • Rack cover can easily be realigned from a traditional lift-off to a hinged rack for one-handed access to tips
  • Front latch closes and locks, ensuring safe and secure storage
  • Space-saving, stackable reload system is environmentally friendly, traceable and easy to use
Envase: Tips are available in racks or in trays for rack reloading. Reload tips require an empty rack.