Borane-tert-butylamine complex ≥95.0% (by titrimetric analysis)

Proveedor: TCI
Peligro

Sinónimos: Boron hydride-tert-butylamine

B1264-25G B1264-100G
TCIAB1264-25GEA 99 EUR
TCIAB1264-25G TCIAB1264-100G
Borane-tert-butylamine complex ≥95.0% (by titrimetric analysis)
Borane-tert-butylamine complex
Formula: C₄H₁₄BN
MW: 86.98 g/mol
Número MDL: MFCD00075635
Núm. CAS: 7337-45-3
EINECS: 230-851-5
UN: 2926
ADR: 4.1 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=242fa3049199247e464e93055d0219ffc6da12ab&vid=242fa3049199247e464e93055d0219ffc6da12ab&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Iodometric Titration) min. 95.0 %
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR