1,1'-Bis(di-tert-butylphosphino)ferrocene ≥95.0% (by titrimetric analysis)

Proveedor: TCI

Sinónimos: 1,1'-Bis(di-t-butylphosphino)ferrocene, 1,1'-Bis[bis(1,1-dimethylethyl)phosphino]ferrocene, DTBPF

B2711-1G B2711-100MG
TCIAB2711-1GEA 81 EUR
TCIAB2711-1G TCIAB2711-100MG
1,1'-Bis(di-tert-butylphosphino)ferrocene ≥95.0% (by titrimetric analysis)
1,1'-Bis(di-tert-butylphosphino)ferrocene
Formula: C₂₆H₄₄FeP₂
MW: 474,42 g/mol
Número MDL: MFCD01630818
Núm. CAS: 84680-95-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7e04f6dc9c1c22c25f8c55aba46d711cc6ad80ba&vid=7e04f6dc9c1c22c25f8c55aba46d711cc6ad80ba&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance Orange to Amber to Dark red powder to crystal
Purity (Chelometric Titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR