4-Oxazolecarboxylic acid ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
O0288-1G
O0288-5G
TCIAO0288-1GEA
51
EUR
TCIAO0288-1G
TCIAO0288-5G
4-Oxazolecarboxylic acid ≥98.0% (by GC, titration analysis)
4-Oxazolecarboxylic acid
Formula:
C₄H₃NO₃ |
Número MDL:
MFCD06797151 Núm. CAS: 23012-13-7 |
|
Resultados test especificaciones
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 140.0 to 144.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...