5-Mercapto-1-methyl-1H-tetrazole ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: 5-Mercapto-1-methyltetrazole

M1081-25G M1081-500G
TCIAM1081-25GEA 51 EUR
TCIAM1081-25G TCIAM1081-500G
5-Mercapto-1-methyl-1H-tetrazole ≥98.0% (by HPLC, titration analysis)
5-Mercapto-1-methyl-1H-tetrazole
Formula: C₂H₄N₄S
Número MDL: MFCD00037317
Núm. CAS: 13183-79-4
EINECS: 236-132-2
UN: 1325
ADR: 4.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f9007778b85bae2c2b32a68b9ffbcf23971dd214&vid=f9007778b85bae2c2b32a68b9ffbcf23971dd214&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Argentmetric Titration) min. 98.0 %
Melting point 124.0 to 129.0 deg-C
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR