N-Methylbenzenesulphonamide ≥98.0% (por GC)
Proveedor: TCI
M0157-25G
M0157-500G
TCIAM0157-25GEA
44
EUR
TCIAM0157-25G
TCIAM0157-500G
N-Methylbenzenesulphonamide ≥98.0% (por GC)
N-Methylbenzenesulphonamide
Formula:
C₇H₉NO₂S MW: 171,21 g/mol |
Número MDL:
MFCD00025023 Núm. CAS: 5183-78-8 EINECS: 225-965-7 |
Resultados test especificaciones
Appearance | White or Colorless to Light yellow powder to lump to clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...