4-(Benzyloxy)phenylboronic acid (contains varying amounts of Anhydride)

Proveedor: TCI
Atención

Sinónimos: 4-Benzyloxybenzeneboronic acid, (4-(Benzyloxy)phenyl)boronic acid

B2145-1G B2145-5G
TCIAB2145-1GEA 24 EUR
TCIAB2145-1G TCIAB2145-5G
4-(Benzyloxy)phenylboronic acid (contains varying amounts of Anhydride)
4-(Benzyloxy)phenylboronic acid
Formula: C₁₃H₁₃BO₃
Número MDL: MFCD01075705
Núm. CAS: 146631-00-7

Order Now

Resultados test especificaciones

Appearance White to Light yellow to Dark green powder to crystal
Purity (Neutralization titration) min. 90.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR