4-Pyridineboronic acid hydrochloride 95%
Proveedor: Thermo Fisher Scientific
|
Atención
|
H52648.03
H52648.06
H52648.03EA
45.6
EUR
H52648.03
H52648.06
4-Pyridineboronic acid hydrochloride 95%
4-Pyridineboronic acid hydrochloride
Purity: 95%
CAS No.: 913835-65-1
Molecular Formula: C5H6BNO2.HCl
Formula Weight: 159.38
Storage Temperature: 2°C to 8°C
Melting Point: 340°
MDL No.: MFCD06201888
Formula:
C₅H₇BClNO₂ Pto. de fusión: 340 °C |
Número MDL:
MFCD06201888 Núm. CAS: 913835-65-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...