Isonicotinic acid-N-oxide ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Atención

I0500-25G I0500-100G
TCIAI0500-25GEA 96 EUR
TCIAI0500-25G TCIAI0500-100G
Isonicotinic acid-N-oxide ≥98.0% (by GC, titration analysis)
Isonicotinic acid-N-oxide
Formula: C₆H₅NO₃
Número MDL: MFCD00006209
Núm. CAS: 13602-12-5
EINECS: 237-086-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c12142656fc7ef674d4baf69af431bacfc341821&vid=c12142656fc7ef674d4baf69af431bacfc341821&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Light yellow to Light red powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Solubility in 1mol/L NaOH almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR