2-Fluoro-4-hydroxybenzoic acid ≥98.0% (by GC, titration analysis)

Proveedor: TCI

F0571-25G F0571-5G
TCIAF0571-25GEA 224 EUR
TCIAF0571-25G TCIAF0571-5G
2-Fluoro-4-hydroxybenzoic acid ≥98.0% (by GC, titration analysis)
2-Fluoro-4-hydroxybenzoic acid
Formula: C₇H₅FO₃
MW: 156,1112 g/mol
Número MDL: MFCD01310985
Núm. CAS: 65145-13-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e4bd246408a5c36d3f3f10046a6f35361ccbebaa&vid=e4bd246408a5c36d3f3f10046a6f35361ccbebaa&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR